Barnstead™ Classic Still Storage Accessories
Thermo Scientific™
Barnstead™ Classic Still Storage Accessories
Complete your distillation system with Thermo Scientific™ Barnstead™ Classic Still Storage Accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
H3111Air Filter and Water Seal
H3240Wall Bracket
Catalog number H3111
Price (INR)
-
Type:
Air Filter and Water Seal
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories complement your current system to create a more complete system. Accessories include mounting options, UV light assembly, Thermo Scientific™ Ventgard™ air filter and water seal. Choose either a floor stand or wall bracket as needed. Maintain sterility in the reservoir with the UV light assembly, which includes housing, on/off switch, 6' cord, lamp and ballast. Protect stored water against airborne particulates, organics and CO2 with Ventgard™ air filter and water seal. Prevent air from entering the reservoir via the condenser atmospheric vent.
Specifications
For Use With (Equipment)Classic Reservoir
TypeAir Filter and Water Seal
Unit SizeEach