Product Image
Invitrogen™
SureCast™ Multi-Use Tool, 12-well
The SureCast™ Multi-Use Tool is a gel knife, gel releaser, and well template guide all in one convenient tool. DesignedRead more
Have Questions?
Catalog NumberQuantity
HC10121 unit
Catalog number HC1012
Price (INR)
1,951.00
온라인 행사
Ends: 30-Sep-2026
3,547.00
Save 1,596.00 (45%)
Each
Quantity:
1 unit
Price (INR)
1,951.00
온라인 행사
Ends: 30-Sep-2026
3,547.00
Save 1,596.00 (45%)
Each
The SureCast™ Multi-Use Tool is a gel knife, gel releaser, and well template guide all in one convenient tool. Designed for use with the SureCast™ Handcast System, this innovative guide outlines the wells for easy visualization and loading of samples into the gel. After electrophoresis, the tool also allows you to easily separate the two glass plates, cut the gel foot, and release the gel.
Includes:
1 SureCast 12-well Multi-Use Tool
Related products:
SureCast™ Handcast Station
SureCast™ Glass Plate Set
SureCast™ Gel Spacers
SureCast™ Sealing Pads
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 unit
Shipping ConditionRoom Temperature
For Use With (Equipment)SureCast Vertical Gel System
Gel SizeMini
Product LineSureCast
Type12-well Multi-Use Tool
Unit SizeEach