imPULSE™ Single-Use Mixer BioProcess Container (BPC)
Thermo Scientific™

imPULSE™ Single-Use Mixer BioProcess Container (BPC)

The Thermo Scientific imPULSE Single-Use Mixer (S.U.M.) BioProcess Container (BPC) is a 3D system composed of ASI 26/77 polyethylene film.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
HM00293I
also known as HM00293-I
2000 L
HM00285I
also known as HM00285-I
30 L
HM00287I
also known as HM00287-I
100 L
HM00288I
also known as HM00288-I
250 L
HM00289I
also known as HM00289-I
500 L
HM00291I
also known as HM00291-I
1000 L
HM00294I
also known as HM00294-I
3000 L
HM00295I
also known as HM00295-I
5000 L
Catalog number HM00293I
also known as HM00293-I
Price (INR)
-
Capacity (Metric):
2000 L
The Thermo Scientific imPULSE Single-Use Mixer (S.U.M.) BioProcess Container (BPC) is a 3D system composed of ASI 26/77 polyethylene film. The innovative disc design and the hexagonal shape of the disc and vessel are at the core of the quick and thorough mixing capabilities of the imPULSE mixer. ImPULSE BPCs help to eliminate post-use cleaning steps required with reusable containers, reduce cross-contamination risks, and are designed to fit the full range of support containers, both square and cylindrical.

Applications:
• Media preparation
• Final formulation steps
• Buffer preparation
• Use as a harvest vessel
• Pooling and liquid transfer
• Product suspension
• Mixing and storage of multiple batches
• Viral inactivation

Customize standard BPC systems for optimized single-use technology
Choose from one of the industry’s largest component libraries with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, and steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade and vent filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Description2,000 L imPULSE Single-Use Mixer (S.U.M.) BioProcess Container (BPC), ASI 26/77 film
Film DescriptionASI™ 26/77
For Use With (Equipment)2000 L imPULSE Single-Use Mixer (S.U.M.)
Outer PackagingDurable cardboard carton
Product LineimPULSE
Protective Inner Packaging3 PE outer layers: supplied flat-packed with two polyethylene outer layers and a box liner cable tied shut
Quantity1 Ea.
Tubing Material Line 11 inlet line: 1.5 in. tri-clover, 0.75 x 1.125 in. ID x OD
Tubing Material Line 21 inflate/vent line: 1.5 in. tri-clover, 0.75 x 1.125 in. ID x OD
Tubing Material Line 31 pressure sensing line: PendoTECH Pressure Sensor, 0.75 in. tri-clover, 0.5 x 0.75 in. ID x OD
Tubing Material Line 41 powder port: 3 in. tri-clover
Tubing Material Line 54 side probe ports: 0.5 in. AseptiQuikTM G Connector
Tubing Material Line 62 outlet/recirculation lines: MPU body with plug, 0.75 x 1.125 in. ID x OD
Capacity (Metric)2000 L
Tubing StyleSilicone
Unit Size2,000 L