Product Image
Thermo Scientific™
Oxoid™ Anaerobic System Schrader Valve, Lid, with clamp and screw
The Oxoid Anaerobic System Schrader Valve is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Have Questions?
Catalog NumberQuantity
HP0015A2/Pk.
Catalog number HP0015A
Price (INR)
-
Quantity:
2/Pk.
Oxoid Anaerobic System Schrader Valve is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Specifications
IncludesLid, complete with clamp and screw
Quantity2/Pk.
Product LineThermo Scientific Oxoid
TypeAnaerobic System Schrader Valve
Unit SizeEach