Oxoid™ Cartridge Container With Desiccant
Thermo Scientific™
Oxoid™ Cartridge Container With Desiccant
Maintain a low moisture environment while safely and conveniently storing unopened Antimicrobial Susceptibility Testing discs.
Have Questions?
Catalog NumberQuantity
HP0055A1/Pk.
Catalog number HP0055A
Price (INR)
-
Quantity:
1/Pk.
Maintain a low moisture environment while safely and conveniently storing unopened Antimicrobial Susceptibility Testing discs in the Thermo Scientific™ Oxoid™ Cartridge Container With Desiccant.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionOxoid Cartridge Container With Desiccant
Quantity1/Pk.
Product TypeCartridge Container with Desiccant
Unit SizeEach