Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Thermo Scientific™
Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)
HR509X20A70
also known as HR509X20A-70
Cryogenic Box, 81-AmpuleFor: Locator 4 Plus
HR509X19ACryogenic Box, 81-AmpuleLocator Jr.
HR509X19A70
also known as HR509X19A-70
Cryogenic Box, 81-AmpuleFor Locator Jr. Plus
HR509X20ACryogenic Box, 81-AmpuleFor Locator 4
Catalog number HR509X20A70
also known as HR509X20A-70
Price (INR)
-
Holds:
Cryogenic Box, 81-Ampule
For Use With (Equipment):
For: Locator 4 Plus
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
  • Designed for long-term storage
  • Features an indexed frame and box organizational system
  • Unique hanging rack promotes easy sample retrieval
  • Suited for both manual and computerized inventory record-keeping methods
Specifications
For Use With (Equipment)For: Locator 4 Plus
HoldsCryogenic Box, 81-Ampule
Length (English)5.25 in.
Width (English)5.25 in.
ColorGray
TypeLocator™ Plus Cryogenic Rack
Unit SizeEach