Search
| Catalog Number | Throughput |
|---|---|
| HYB18ABOFX | 4 Enrichments |
| HYB18ABOFX8 | 8 Enrichments |
Simplified workflow
Flexible, modular genotyping
Quality results
Improved data quality
Expand Understanding
Box 1, Box 3 and the oligonucleotide probe mix can be safely stored at -20°C and Box 2 can be safely stored at 4°C.