One Lambda™ HybriType™ HLA Plus 18 Loci Flex Kit, 4 Enrichments
Thermo Scientific™
One Lambda™ HybriType™ HLA Plus 18 Loci Flex Kit, 4 Enrichments
One Lambda™ HybriType™ HLA Plus 18 Loci Flex Kit – 4 Enrichments, provides additional probes for the targeted enrichment of 18 HLA loci (classical and non-classical) and MIC genes for use with One Lambda™ HybriType™ HLA Plus Typing Flex Kit – 96
Have Questions?
Catalog NumberQuantity
HYBHLA3FX4 Enrichments
Catalog number HYBHLA3FX
Price (INR)
-
Quantity:
4 Enrichments
Additional enrichment reagents conveniently support continued HLA and MICA/B sequencing when more than four runs are needed to process 96 samples and enrichment reagents from the One Lambda™ HybriType™ HLA Plus Typing Flex Kit are depleted.
Flexible Sample Management
  • Reduces the need to batch the same number of samples in every run of the One Lambda™ HybriType™ HLA Plus Typing Flex Kit – 96
Reduce delay:
  • Batch samples according to actual sample throughput not prescriptive requirements
Reduce costs
  • Minimize scrap, when the One Lambda™ HybriType™ HLA Plus Typing Flex Kit – 96 enrichment reagents are prematurely depleted: using the remaining reagents with the ancillary One Lambda™ HybriType™ HLA Plus 18 Loci Flex Kit – 4 Enrichments (HYBHLA3FX) kit to continue sample typing
Specifications
Product TypeHLA Typing Ancillary
Quantity4 Enrichments
Regulatory StatusRUO
FormatAncillary Reagents
Sequencing TypeShort Read Sequencing
Unit SizeEach
Contents & Storage
  • Qty:1; Contains 9 reagents designed for processing 4 pooled libraries, optimized for hybrid capture enrichment of 18 loci (11 classical and 7 non-classical)
  • The box can be safely stored at -20°C.