iGlow™ Gel Documentation System
Invitrogen™
iGlow™ Gel Documentation System
The Invitrogen iGlow Gel Documentation System is a flexible and intuitive imaging platform that supports fast and easy nucleic acid and protein gel imaging across a diverse range of gel formats, offering researchers flexibility in sample preparation and simplicity in imaging.
Have Questions?
Catalog NumberVoltage
IGLOW220200/240 V
Catalog number IGLOW220
Price (INR)
1,305,480.00
Each
Voltage:
200/240 V
Price (INR)
1,305,480.00
Each
The Invitrogen iGlow Gel Documentation System is a flexible and intuitive imaging platform for nucleic acid and protein gels. Featuring 'one touch capture' functionality, the image acquisition process takes mere seconds to go from sample insertion to image acquisition. Automatic tray recognition simplifies the imaging process by sensing which sample tray is inserted and only activating the corresponding imaging mode. Featuring a built-in 12 inch touchscreen interface, three distinct LED transilluminator light sources (blue, UV, and white), and a sCMOS camera for high-quality image capture, the iGlow enables precision imaging for a wide range of nucleic acid and protein gel samples and stains.
Product Features:
  • One-Touch Image Capture System: Streamlined operation with single-touch activation enables rapid image acquisition and enhanced user efficiency for high-throughput laboratory workflows
  • Automated Tray Recognition Technology: Intelligent detection system automatically identifies sample trays, removing manual steps and reducing potential for user error.
  • Integrated Touchscreen Interface: Built-in display provides intuitive navigation and system control, enabling user control without the need for external computing devices
  • High Resolution 5-MP CMOS Camera: Powerful imaging sensor enables publication-quality image capture with superior clarity and detail suitable for scientific documentation and analysis
  • Multi-Source Gel Excision Capability: Supports precision gel excision workflows utilizing white, blue and UV light sources for versatile sample preparation applications
  • Integrated Safety Detection System: Automated safety screen monitoring ensures UV shield remains in place during gel excision procedures, preventing potential user exposure
  • Comprehensive Data Transfer Options: Multiple connectivity solutions including Ethernet, Wi-Fi, and USB interfaces enable flexible data management and easy integration with network infrastructures
System Specifications and Operational Overview
Advanced Imaging Technology with Integrated Computing Platform
The system incorporates a sophisticated Windows-based computer platform that cleanly integrates hardware and software components. At the core of the imaging capabilities is a high-resolution 5-megapixel scientific CMOS detector specifically designed for precision image capture. Advanced sensor technology provides automated recognition capabilities of sample trays and the UV safety shield, promoting operational safety and workflow efficiency.
Intuitive Automated Imaging Workflow
Operation of the system follows an intuitive workflow designed to maximize efficiency while maintaining scientific rigor. Users begin by opening the sample drawer and positioning their specimen on one of the imaging trays. The onboard software activates only the imaging mode that matches the inserted sample tray, simplifying the user experience. When capturing an image the system automatically detects sample intensity levels, calculates an exposure time and captures an image. Automated features reduce guesswork and promote consistent, high-quality results across diverse sample types.
Comprehensive Data Management and Image Processing Capabilities
The iGlow has onboard image editing capabilities, allowing users to perform basic image adjustments directly on the imager. Following image acquisition, users can export captured data to network locations or USB storage devices. These images are fully compatible with the iBright Analysis Software (iBA), enabling users to do advanced image analysis, add annotations and create custom data reports on Mac, PC or in the Cloud.
Complete System Package with Extended Support Coverage
The system includes all essential components required for operation. All imaging trays, the UV safety shield, and a convenient tray rack, are included with the imager. Furthermore, the system is backed by a comprehensive two-year warranty, providing extended coverage and peace of mind for laboratory operations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Camera TypesCMOS
Capacity400GB Hard drive
DimensionsWxDxH: 15 x 18 x 16 in. (38 x 45 x 41 cm)
Field5.5 x 8.3 in. (14 x 21 cm) Field of View
FormatImage formats: .iba (proprietary), TIFF, JPG
IncludesWhite, Blue and UV Imaging Trays, Tray Rack, UV Safety Shield
ModeliGlow
SoftwareiBright Analysis Software
TransilluminatorWhite, Blue and UV LED Matrix
UV MonitoringAutomated Safety Shutoff
Wavelength590 nm
Weight (English)48.5 lb.
Weight (Metric)22 kg
Display TypeLCD Touchscreen
Filter TypeEmission Filter
For Use With (Application)Gel Documentation
Pixel Size5 MP
Product LineiGlow
Product TypeGel Documentation System
Voltage200/240 V
Unit SizeEach
Frequently asked questions (FAQs)
Can the iGlow Gel Documentation System be used in shared laboratory environments?
Yes. The iGlow Gel Documentation System includes One Touch Capture workflows, method saving capabilities, and automated tray recognition features that can help support consistent operation across multiple users.
What is the maximum gel size supported by the iGlow Gel Documentation System?
The iGlow Gel Documentation System supports gels up to approximately 20 × 13 cm in size and can image up to four mini gels or two midi gels simultaneously.
When should I choose an iGlow Gel Documentation System instead of an iBright Imaging System?
The iGlow Gel Documentation System is designed specifically for gel imaging workflows. Laboratories that require chemiluminescence imaging, western blot imaging, or more advanced imaging applications may wish to consider an iBright Imaging System.
Which types of gel imaging workflows and applications is the iGlow Gel Documentation System designed to support?
The iGlow Gel Documentation System is designed for routine imaging of nucleic acid and protein gels, including fluorescently stained DNA gels, protein gels, and colorimetric gels. It is well suited for laboratories that require fast, simple gel imaging workflows without chemiluminescence imaging capabilities.
Does the iGlow Gel Documentation System allow users to manually control imaging parameters?
Yes. Users can adjust imaging parameters including field of view, exposure time, and illumination intensity (UV and blue illumination). Imaging settings can also be saved as methods for future use.