imPULSE™ Single-Use Mixer (S.U.M.)
Thermo Scientific™

imPULSE™ Single-Use Mixer (S.U.M.)

Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
IM001009002
also known as IM00100.9002
100 L
IM000309001
also known as IM00030.9001
30 L
IM002509003
also known as IM00250.9003
250 L
IM005009004
also known as IM00500.9004
500 L
IM010009005
also known as IM01000.9005
1000 L
IM020009006
also known as IM02000.9006
2000 L
IM030009007
also known as IM03000.9007
3000 L
IM050009008
also known as IM05000.9008
5000 L
Catalog number IM001009002
also known as IM00100.9002
Price (INR)
-
Capacity (Metric):
100 L
The Thermo Scientific imPULSE Single-Use Mixer (S.U.M.) can be used for many bioprocess mixing applications. The imPULSE S.U.M. design features include innovative disc-mixing technology, configurable high-end controls, and monitors to fit specific process requirements. These features enable uniform, superior mixing that is scalable from 30 L to 5,000 L. The touchscreen console for the imPULSE S.U.M. provides unsurpassed integrated sensor monitoring and pump control for pH and saline titration and for automatic fill and harvest of the S.U.M.

imPULSE S.U.M features include:
• Touchscreen console provides ease of use with multi-functional capabilities to monitor and control mixing parameters
• Integrated rolling diaphragm that provides the pumping action to the mixing disc; the diaphragm will not abrade the surfaces or produce particulate
• Mixing tank jacket and insulation
• Load cell weighing systems enable accurate batch weight monitoring
• Auto inflate and vent control options
• Adjustable powder hanger for 1,000 L and higher mixers that fits 1, 5, and 25 kg Powdertainer BPCs
• Open cart frame for easier cleaning

Applications:
• Media preparation
• Final formulation steps
• Buffer preparation
• Use as a harvest vessel
• Pooling and liquid transfer
• Product suspension
• Mixing and storage of multiple batches
• Viral inactivation

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Description100 L imPULSE Single-Use Mixer (S.U.M.)
Power RequirementsAC motor module: Powered by the TSC* TSC: 15 A, 100–120/208–240 VAC, 50–60 Hz
Quantity1 Ea.
Capacity (Metric)100 L
Unit SizeEach