ExcelExchange Module for GRAMS Suite Spectroscopy Software
Thermo Scientific™
ExcelExchange Module for GRAMS Suite Spectroscopy Software
Easily share data between Thermo Scientific™ GRAMS™ Suite Spectroscopy Software applications and Microsoft Excel™ spreadsheets.
Have Questions?
Catalog NumberFor Use With (Equipment)
INF-15020GRAMS/AI
Catalog number INF-15020
Price (INR)
-
For Use With (Equipment):
GRAMS/AI
Use the Thermo Scientific™ ExcelExchange module to easily share data between Thermo Scientific™ GRAMS™ Suite Spectroscopy Software applications and Microsoft Excel™ spreadsheets. The ExcelExchange module is an effective way to import data stored in Excel spreadsheets or complex ASCII data sets into Thermo Scientific™ GRAM/AI™ Spectroscopy Software, and to export multidimensional spectral data into Excel spreadsheets for custom calculations.
Exchanging data between the GRAMS format and Excel
  • Converts Thermo Scientific™ GRAMS/AI™ SPC format spectral data files to a Microsoft Excel worksheet
  • Transforms Microsoft Excel worksheets into 2D or 3D GRAMS SPC files
  • Imports data stored in ASCII, CSV, or text formats or reports
  • Modify the data within ExcelExchange and easily select the exact data for conversion
Specifications
DescriptionExcelExchange
For Use With (Equipment)GRAMS/AI
TypeModule
Unit SizeEach