ARL iSpark Plus Inclusion Analyzer
ARL iSpark Plus Inclusion Analyzer
ARL iSpark Plus Inclusion Analyzer
ARL iSpark Plus Inclusion Analyzer
Thermo Scientific™
ARL iSpark Plus Inclusion Analyzer
Are you facing quality issues in your steel products, or in your process, caused by inclusions? The ARL iSpark 8860 Inclusion Analyzer performs inclusion analysis simultaneously with elemental analysis, offering you unique solutions to check and control inclusions
Have Questions?
Catalog NumberType
IQLAAHGABDFAOOMBODInclusion Analyzer
Catalog number IQLAAHGABDFAOOMBOD
Price (INR)
-
Type:
Inclusion Analyzer
The ARL iSpark Plus Inclusion Analyzer with Spark-DAT has unparalleled speed for inclusion analysis in steel. Thanks to Spark-DAT methods, inclusion data are obtained at the same time as the elemental concentrations. It is possible to evaluate inclusions in all the samples analyzed in concentration with the OES spectrometer, up to several hundred samples per day. This provides fast and efficient solutions to avoid detrimental effects of inclusions on steel properties and prevent costly issues like nozzle clogging during the continuous casting process.
Key Features
Non-metallic inclusions are always present in steel, to an extent that depends on the steel grade and the production process. Since inclusions strongly affect steel properties and smooth running of the production process, they must be controlled. The Thermo Scientific™ ARL iSpark™ Plus Inclusion Analyzer delivers combined elemental concentrations and inclusion data in the same analysis, making control of inclusions possible in almost real time during steel manufacturing, in up to hundreds of steel production samples per day.
For more than twenty years, our company has been a pioneer in the ultra-fast analysis of non-metallic micro-inclusions by spark OES. Thermo Scientific’s Spark-DAT inclusion analysis available today with the ARL iSpark Plus Inclusion Analyzer builds on this long experience. The ARL iSpark Plus Inclusion Analyzer comes as a comprehensive package including all the features needed for a thorough and rapid elemental analysis of steel combined with inclusion analysis. It will help guarantee optimal inclusion content in the steel products, preventing occurrences of costly process issues like nozzle clogging, and minimizing claims and returns of products by customers.
The Thermo Scientific ARL iSpark Plus Inclusion Analyzer sports powerful features:
  • ARL iSpark Plus, the trusted standard in spark OES, fully equipped for top performance elemental analysis and most advanced inclusion analysis in steel
  • 1m vacuum Paschen-Runge optics with PMT detectors
  • Full elemental coverage, including gas elements
  • Ultra-fast inclusion analysis, combined with elemental analysis
  • Ability to determine inclusion type, number, size, concentration, volume fraction and more
  • All inclusion analysis software tools, including Spark Explorer, Inclusions Report and SPC
  • Customization of the inclusion analysis method and training with Thermo Fisher Scientific experts
  • Perfectly adapted for ARL iSpark Plus automated with ARL SMS (Sample Manipulation System)
Specifications
Frequency50/60 Hz
Height (Metric) Exterior1200 mm
Weight (Metric)Approximately 500 kg
Width (Metric) Exterior860 mm
Detector TypePhotomultiplier Tubes (PMT)
For Use With (Application)Steel Making
TypeInclusion Analyzer
Unit SizeEach