Orbitrap™ Isora™ Mass Spectrometers
Orbitrap™ Isora™ Mass Spectrometers
Orbitrap™ Isora™ Mass Spectrometers
Thermo Scientific™

Orbitrap™ Isora™ Mass Spectrometers

Measure isotope and isotopologue ratios directly from intact molecular ions using the electrospray ionization and high-resolution accurate mass detection of the Thermo Scientific™ Orbitrap™ Isora™ Mass Spectrometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelMax. Resolution
IQLAAMGAATFASMMBQHOrbitrap Isora Pro Mass SpectrometerUp to 480,000 at 200 m/z
IQLAAMGAATFASMMBQIOrbitrap Isora Mass SpectrometerUp to 240,000 at 200 m/z
Catalog number IQLAAMGAATFASMMBQH
Price (INR)
-
Model:
Orbitrap Isora Pro Mass Spectrometer
Max. Resolution:
Up to 480,000 at 200 m/z

Extend your capabilities for isotope ratio analysis across applications including environmental research, geochemistry, metabolism studies, and food authenticity. The purchase of an Orbitrap Isora Mass Spectrometer includes:

  • One Orbitrap Isora Mass Spectrometer
  • Dedicated Thermo Scientific™ Vanquish™ Duo UHPLC system configuration
  • Thermo Scientific™ Isotope Discoverer™ Software

to help offer a straightforward path from measurement to final isotope ratio data.

Reveal isotope ratios in molecular context

Innovative Orbitrap-based isotopologue analysis opens access to additional dimensions of isotope ratio information, enabling deeper, more comprehensive studies of isotopic composition across molecular species.

Expand your analytical capabilities by analyzing:

  • Isotope ratios at natural abundance: simultaneous acquisition of major and selected minor isotopologues for high precision isotopic analysis of elements such as C, N, O, H and S
  • Position-specific isotope ratios: controlled fragmentation of organic molecular species to measure isotope ratios from specific functional groups or molecular fragments
  • Clumped isotopes: measurements of isotopologues with multiple rare isotope substitutions

Integrated workflow from sample to result

Designed to simplify complex isotope workflows, the Orbitrap Isora MS solution helps laboratories move efficiently from measurement to evaluated results, even with large, information-rich datasets. Included with your purchase:

  • Orbitrap Isora MS with purpose-built control software for method development and acquisition setup
  • Vanquish Duo UHPLC System supports sample introduction and rapid sample/reference setup for accuracy of isotopic data
  • Isotope Discoverer software for integrated data processing, visualization and reporting

Isotope Discoverer software for streamlined data processing

Designed to make isotope data processing more efficient, traceable, and easier to navigate, Isotope Discoverer Software brings key processing steps into one environment, transforming complex high-resolution data into final isotope ratio results with greater speed and clarity.

Key capabilities include:

  • Extract isotope ratio information directly from high-resolution raw data
  • Filter, summarize, and correct data within a structured workflow
  • Calibrate results to generate final delta values
  • Visualize intermediate and final results for review and troubleshooting
  • Export processed data and reports for further analysis and documentation
Specifications
Dimensions (D x W x H)30 x 21 x 27.6 in. (763 x 535 x 703 mm)
Dynamic Range5 orders of magnitude intrascan dynamic range
FragmentationHigher Collisional Dissociation (HCD)
Mass AccuracyUnder defined conditions: External calibration achieves <3 ppm RMS; Internal calibration achieves <1 ppm RMS
Mass StabilityUnder defined conditions: External calibration achieves <3 ppm mass drift over 24 hours; Internal calibration achieves <1 ppm mass drift over 24 hours; EASY-IC (scan-to-scan lock mass correction) achieves <1 ppm mass drift for at least 5 days; 1-point mass calibration achieves <3 ppm mass drift over 4 weeks.
ModelOrbitrap Isora Pro Mass Spectrometer
Max. ResolutionUp to 480,000 at 200 m/z
TypeOrbitrap IRMS
Unit SizeEach

Frequently asked questions (FAQs)

Is there training and support available for the Thermo Scientific Orbitrap Isora Mass Spectrometer?

Yes. Thermo Fisher Scientific provides support for the Thermo Scientific Orbitrap Isora Mass Spectrometer through its global service and application support organization. In addition to instrument installation and familiarization, the integrated workflow is designed to simplify adoption by including purpose-built control software for method development and acquisition, the dedicated Thermo Scientific Vanquish Duo UHPLC system, and Thermo Scientific Isotope Discoverer Software for data processing, visualization, and reporting. Customers also have access to Thermo Fisher technical documentation, application resources, and optional training programs to help users become proficient in instrument operation, workflow development, and isotope ratio data analysis.

Is the purchase of the Thermo Scientific Vanquish required for the Thermo Scientific Orbitrap Isora Mass Spectrometer?

The Thermo Scientific Orbitrap Isora Mass Spectrometer is offered as an integrated isotope ratio analysis solution: the standard configuration includes one Orbitrap Isora Mass Spectrometer, a dedicated Thermo Scientific Vanquish Duo UHPLC system configuration, and Thermo Scientific Isotope Discoverer Software. The dedicated Vanquish Duo UHPLC system is an important part of the workflow, providing sample introduction and rapid sample/reference switching to support accurate isotope ratio measurements. This integrated configuration is designed to provide a streamlined path from sample measurement through data processing and final isotope ratio results.

How can the Thermo Scientific Orbitrap Isora Mass Spectrometer complement existing isotope ratio workflows in my laboratory?

The Thermo Scientific Orbitrap Isora Mass Spectrometer complements classical magnetic sector IRMS workflows by enabling isotope ratio measurements directly from intact molecular ions using electrospray ionization and Orbitrap HRAM detection. Rather than requiring conversion of samples into simple gases, it preserves molecular information while allowing analysis of isotope ratios of individual analytes at natural abundance and low-level isotopic enrichment, position-specific isotope analysis, and multiply substituted (clumped) isotopes within a single platform. Depending on the application, the Orbitrap Isora MS can either complement classical IRMS by providing additional molecular context or serve as an alternative workflow for analyses requiring intact molecular isotopic information with simplified sample preparation and with high sensitivity.

What should I consider when choosing between the Thermo Scientific Orbitrap Isora MS and Thermo Scientific Orbitrap Isora Pro MS models?

The primary consideration is the level of analytical performance required. The Thermo Scientific Orbitrap Isora MS offers maximum resolution up to 240,000 (at m/z 200), making it suitable for many isotope ratio applications. The Thermo Scientific Orbitrap Isora Pro MS doubles the maximum resolution up to 480,000 (at m/z 200), providing greater resolving capability for complex isotopologue measurements. Orbitrap Isora Pro MS has additional features that support users beyond conventional HRAM detection and quantification of compounds and isotope-focused analysis, such as: Peptide operation mode, Thermo Scientific TMTpro 35-plex, Bent trap pre-accumulation, and Hybrid-DIA with adaptive RT routine. Both models include the same integrated workflow components—Thermo Scientific Vanquish Duo UHPLC System and Thermo Scientific Isotope Discoverer Software—providing a consistent sample-to-result experience.

Which laboratories are well-suited for the Thermo Scientific Orbitrap Isora Mass Spectrometer?

The Thermo Scientific Orbitrap Isora Mass Spectrometer is well suited for laboratories tackling applications in environmental research, geochemistry, metabolism studies, fluxomic research and food authenticity that are seeking comprehensive isotopic characterization of samples. By combining molecular-level information with isotope ratios, Orbitrap Isora MS can help to address new analytical questions with greater depth and specificity, supporting the following investigations:

  1. Origin (Where did this molecule come from?)
  2. Formation (How was it synthesized?)
  3. Transformation (What chemical or biological processes has it undergone?)
  4. Identity (Is it authentic, adulterated, or distinguishable from similar materials?)
  5. Mechanism (Which atoms or pathways are involved in its formation or metabolism?)