Product Image
Invitrogen™

iPrep™ Card: gDNA Tissue

iPrep™ Card: gDNA Tissue
Have Questions?
Catalog NumberQuantity
IS100131 card
Catalog number IS10013
Price (INR)
69,281.00
線上優惠
Ends: 30-Sep-2026
125,965.00
Save 56,684.00 (45%)
Each
Quantity:
1 card
Price (INR)
69,281.00
線上優惠
Ends: 30-Sep-2026
125,965.00
Save 56,684.00 (45%)
Each
The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.
The following iPrep™ Protocol Cards are available from Invitrogen:
- iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
-iPrep™ Tissue Card for use with the iPrep ChargeSwitch™ Tissue Kit
- iPrep™ Blood Card for use with the iPrep PureLink Blood kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)iPrep™
Product LineiPrep
Quantity1 card
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Do you still offer the iPrep Purification Instrument?

The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.