The iPrep™ Viral DNA/RNA protocol Card is for use with the iPrep™ PureLink™ Virus Kit. The iPrep™ Protocol Card isRead more
Have Questions?
Catalog Number
Quantity
IS10016
1 card
Catalog number IS10016
Price (INR)
123,251.00
Each
Quantity:
1 card
Price (INR)
123,251.00
Each
The iPrep™ Viral DNA/RNA protocol Card is for use with the iPrep™ PureLink™ Virus Kit. The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time. The following iPrep™ Protocol Cards are available from Invitrogen: • iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit • iPrep™ Tissue Card for use with the iPrep™ ChargeSwitch™ Tissue Kit • iPrep™ Blood Card for use with the iPrep™ PureLink™ Blood kit • iPrep™ Viral DNA/RNA Card for use with the iPrep™ PureLink™ Virus Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 card
FormatCard
Product LineiPrep
Unit SizeEach
Frequently asked questions (FAQs)
Do you still offer the iPrep Purification Instrument?
The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.