Attune™ Wash Solution

Attune™ Wash Solution

Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
J249741 L
A24974250 mL
Catalog number J24974
Price (INR)
15,289.00
Each
Quantity:
1 L
Price (INR)
15,289.00
Each

Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.

Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.

Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.

High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.

Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.

Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.

Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.

High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Attune™ Acoustic Focusing Cytometer
Product LineAttune
Quantity1 L
Product TypeWash Solution
Unit SizeEach
Contents & Storage
Store at room temperature.