RevertAid™ First Strand cDNA Synthesis Kit, with DNase I
RevertAid™ First Strand cDNA Synthesis Kit, with DNase I
RevertAid™ First Strand cDNA Synthesis Kit, with DNase I
Thermo Scientific™

RevertAid™ First Strand cDNA Synthesis Kit, with DNase I

The Thermo Scientific RevertAid First Strand cDNA Synthesis Kit with DNase I is a complete system for efficient synthesis ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K16225100 Reactions
K1621520 Reactions
Catalog number K16225
Price (INR)
53,770.00
Each
No. of Reactions:
100 Reactions
Price (INR)
53,770.00
Each

The Thermo Scientific RevertAid First Strand cDNA Synthesis Kit with DNase I is a complete system for efficient synthesis of first strand cDNA from RNA templates. This kit includes recombinant endonuclease (DNase I) to remove contaminating genomic DNA from RNA preps.

For reverse transcription the kit uses RevertAid Reverse Transcriptase (RT), a recombinant M-MuLV RT that maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA templates from degradation.

The RevertAid First Strand cDNA Synthesis Kit is supplied with both oligo(dT)18 and random hexamer primers. The oligo(dT)18 primer anneals selectively to the poly(A) tail of mRNA. Random hexamer primers do not require a poly(A) tail, so can be used for transcription of the 5'-end regions of mRNA or cDNA synthesis of RNA species lacking a poly(A) tail (e.g., microRNAs). Gene-specific primers may also be used with the kit.

Features of the RevertAid First Strand cDNA Synthesis Kit with DNase I include:
• Integrated genomic DNA removal step
• Full-length first-strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit—all components for the RT reaction are included

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
No. of Reactions100 Reactions
Optimal Reaction Temperature42°C
Quantity100 reactions
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseRevertAid
Ribonuclease H ActivityYes
Size (Final Product)Up to 13 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)Real Time PCR (qPCR), RT-PCR
Reaction Speed60 min.
Unit SizeEach
Contents & Storage

1 box containing:
• RevertAid Reverse Transcriptase, 1 x 120 μL (200 U/μL)
• RiboLock RNase Inhibitor, 1 x 120 μL (20 U/μL)
• 5X Reaction Buffer, 1 x 500 μL
• dNTP Mix, 1 x 250 μL (10 mM)
• Oligo(dT)18 Primer, 1 x 120 μL (100 μM)
• Random Hexamer Primer, 1 x 120 μL (100 μM)
• Control GAPDH RNA, 1 x 20 μL (0.05 μg/μL)
• Forward GAPDH Primer, 1 x 20 μL (10 μM)
• Reverse GAPDH Primer, 1 x 20 μL (10 μM)
• Nuclease-free water, 2 x 1.25 mL

5 boxes, each containing:
• DNase I, RNase-free, 1 x 20 μL (1 U/μL)
• 10X Reaction Buffer with MgCl2 for DNase I, 1 x 1.25 mL
• EDTA, 1 x 1 mL (50 mM)
• 10X Reaction Buffer without MnCl2, 1 x 1 mL
• MnCl2, 1 x 1 mL (100 mM)

Store at –20°C.