Product Image
Thermo Scientific™
pENTR™/D-TOPO™ Cloning Kit
The pENTR™/D-TOPO™ Cloning Kits utilize a highly efficient, 5-minute cloning strategy (“TOPO™ Cloning”) to directionally clone a blunt-end PCR productRead more
Have Questions?
Catalog NumberQuantity
K240020SP20 Reactions
Catalog number K240020SP
Price (INR)
26,408.00
線上優惠
Ends: 30-Sep-2026
48,014.00
Save 21,606.00 (45%)
Each
Quantity:
20 Reactions
Price (INR)
26,408.00
線上優惠
Ends: 30-Sep-2026
48,014.00
Save 21,606.00 (45%)
Each
The pENTR™/D-TOPO™ Cloning Kits utilize a highly efficient, 5-minute cloning strategy (“TOPO™ Cloning”) to directionally clone a blunt-end PCR product into a vector for entry into the Gateway™ System or the MultiSite Gateway™ System. Blunt-end PCR products clone directionally at greater than 90% efficiency, with no ligase, post-PCR procedures, or restriction enzymes required.
The kit comes with everything necessary to clone and select your PCR amplified gene of interest:
Gateway™ System Ready—Rapidly shuttle cloned genes between multiple vector systems
Fast and Easy—Go from PCR to Gateway™ Entry clone in just 3 steps and as little as ∼5 minutes hands on time
Efficient—Achieve over 90% clones with correct insert in the right direction
Proven—Reliable performance for over a decade
pENTR™ /D-TOPO™ Cloning Kits Overview
VECTORpENTR™ /D-TOPO™ VectorDirectional cloning vector for entry to the Gateway System
CLONING METHODDirectional TOPO™ CloningTopoisomerase I based ∼5 minute directional ligation of blunt-end proofreading polymerase-amplified PCR products to the vector
COMPETENT CELLSTwo OptionsChoose from kits with either high efficiency, or fast growing competent cells
Simple Access to the Gateway™ System
For access to the Gateway™ System, just PCR amplify your gene of interest and add the product straight to the provided topoisomerase charged pENTR™/D-TOPO™ vector, incubate 5 minutes, and transform the provided competent E. coli cells. The resulting attL containing Gateway™ Entry clones are ready for efficient recombination with your choice of Gateway™ Destination vectors.
Optimized pENTR™/D-TOPO™ Vector
The pENTR™/D-TOPO™ vector (Figure 1) includes M13 and T7 primer sequencing sites and attL recombination sites flanking the PCR product insertion site. This allows clones to be easily sequence verified and recombined into your choice of attR containing Gateway™ destination vectors. A Kanamycin resistance gene and a pUC origin are used for selection and high-copy propagation in E. coli.
Simplified Directional Cloning
With Directional TOPO™ cloning technology there is no need for PCR clean up, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase charged vector, incubate 5 minutes, transform, and obtain up to 90% directionally inserted clones. A four base overhang on the vector pairs with a four base sequence designed into the forward primer used in your PCR reaction to provide directionality to the topoisomerase ligation reaction (Figure 2).
The Power of Gateway™ Recombination Cloning Technology
Gateway™ recombination cloning technology circumvents the limitations of restriction mediated cloning, enabling you to access virtually any expression system in a simple one hour, 99%-efficient and reversible, Gateway™ recombination reaction. The ability to move the same sequence of DNA between different vectors without using restriction enzymes, ligase, subcloning steps, screening of countless colonies, or re-sequencing will help save you time, money, and effort.
Leading Cloning Technologies
When it comes to cloning, TOPO™ cloning technology and Gateway™ recombination cloning technology have been a reliable partner for thousands of scientists for over ten years. Fast, simple to use, and efficient, TOPO™ cloning and Gateway™ recombination allow for rapid cloning and subsequent transferring of genes between a wide assortment of Gateway™ expression vectors.
Specifications
Bacterial or Yeast StrainTOP10
Cell TypeChemically Competent E. coli
Cloning MethodTOPO cloning
For Use With (Application)Cloning
Product TypeCloning Kit
PromoterT7
Quantity20 Reactions
FormatKit
Unit SizeEach
Contents & Storage
pENTR™/D-TOPO™ Cloning Kit contains pENTR/D-TOPO™ vector, dNTPs, salt solution, sterile water, and universal M13 sequencing primers.
Store components at -20°C.