Product Image
Invitrogen™

pENTR™/SD/D-TOPO™ Cloning Kit, with One Shot™ Mach1™-T1R Chemically Competent E. coli

The pENTR™/SD/D-TOPO™ Cloning Kits utilize a highly efficient, 5-minute cloning strategy (“;TOPO™ Cloning”) to directionally clone a blunt-end PCR productRead more
Have Questions?
Catalog NumberQuantity
K26352020 Reactions
Catalog number K263520
Price (INR)
-
Quantity:
20 Reactions
The pENTR™/SD/D-TOPO™ Cloning Kits utilize a highly efficient, 5-minute cloning strategy (“;TOPO™ Cloning”) to directionally clone a blunt-end PCR product into a vector for entry into the Gateway™ System or the MultiSite Gateway™ System. Blunt-end PCR products clone directionally into the pENTR™/SD/D-TOPO™ entry vector at greater than 90% efficiency, and include an upstream Shine-Dalgarno sequence for expression in prokaryotes.

The kit comes with everything necessary to clone and select your PCR amplified gene of interest:

Gateway™ System Ready - Rapidly shuttle cloned genes between multiple vector systems
Fast and Easy - Go from PCR to Gateway™ Entry clone in just 3 steps and as little as ∼5 minutes hands on time
Efficient - Achieve over 90% clones with correct insert in the right direction
Proven - Reliable performance for over a decade

pENTR™/SD/SD D-TOPO™ Cloning Kits Overview

VECTOR

pENTR™ /SD SD/SD D-TOPO™ Vector Directional cloning vector for entry to the Gateway™ System and optimized for prokaryotic expression

CLONING METHOD

Directional TOPO™ Cloning Topoisomerase I based ∼5 minute directional ligation of blunt-end proofreading polymerase-amplified PCR products to the vector

COMPETENT CELLS

Two Options Choose from kits with either high efficiency, or fast growing competent cells
Simple Access to the Gateway™ System
For access to the Gateway™ System, just PCR amplify your gene of interest and add the product straight to the provided topoisomerase charged pENTR™ /SD SD/SD D-TOPO™ vector, incubate 5 minutes, and transform the provided competent E. coli cells. The resulting attL containing Gateway™ Entry clones are ready for efficient recombination with your choice of Gateway™ Destination vectors.

Optimized pENTR™ /SD SD/SD D-TOPO™ Vector
The pENTR™ /SD SD/SD D-TOPO™ vector (Figure 1) includes a T7 gene 10 translational enhancer and a ribosome binding site (RBS) for optimal expression of native protein after recombination with a prokaryotic Gateway™ destination vectors. You can also use the vector for expression in other hosts systems by recombination with different destination vectors. The vector has M13 and T7 primer sequencing sites and attL recombination sites flanking the PCR product insertion site. This allows clones to be easily sequence verified and recombined into your choice of attR containing Gateway™ destination vectors. A Kanamycin resistance gene and a pUC origin are used for selection and high-copy propagation in E. coli.

Simplified Directional Cloning
With Directional TOPO™ cloning technology there is no need for PCR clean up, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase charged vector, incubate 5 minutes, transform, and obtain up to 90% directionally inserted clones. A four base overhang on the vector pairs with a four base sequence designed into the forward primer used in your PCR reaction to provide directionality to the topoisomerase ligation reaction (Figure 2).

The Power of Gateway™ Recombination Cloning Technology
Gateway™ recombination cloning technology circumvents the limitations of restriction mediated cloning, enabling you to access virtually any expression system in a simple one hour, 99%-efficient and reversible, Gateway™ recombination reaction. The ability to move the same sequence of DNA between different vectors without using restriction enzymes, ligase, subcloning steps, screening of countless colonies, or re-sequencing will help save you time, money, and effort.

Leading Cloning Technologies
When it comes to cloning, TOPO™ cloning technology and Gateway™ recombination cloning technology have been a reliable partner for thousands of scientists for over ten years. Fast, simple to use, and efficient, TOPO™ cloning and Gateway™ recombination allow for rapid cloning and subsequent transferring of genes between a wide assortment of Gateway™ expression vectors.

Kit Options
The pENTR™/SD/D-TOPO™ Cloning Kit can be purchased with either TOP10 competent cells for standard cloning or Mach1™-T1R competent cells for fast growth.

For Research Use Only. Not intended for use in animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodDirectional TOPO™
No. of Reactions20 reactions
Product LineOne Shot
Product TypeTOPO Cloning Kit
Quantity20 Reactions
VectorpENTR
Unit SizeEach
Contents & Storage
Each Directional TOPO™ pENTR Cloning Kit contains the appropriate, ready-to-use TOPO™ vector, dNTPs, salt solution, sterile water, universal M13 sequencing primers, TOP10 or Mach1-T1R Chemically E. coli, S.O.C. Medium, and a pUC19 control plasmid. Store Competent E. coli at -80°C. Store all other components at -20°C.

Frequently asked questions (FAQs)

Your Gateway-adapted TOPO vectors are supplied with a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

I am using a Directional TOPO Cloning vector for cloning my PCR product. I have screened a bunch of colonies but they all contain my insert cloned in the reverse direction. How can I solve this problem?

Here are possible causes and suggestions:

- Incorrect PCR primer design: Make sure that the forward PCR primer contains the sequence, CACC, at the 5' end. The 4 nucleotides, CACC, base pair with the overhang sequence, GTGG, in the Directional TOPO vector.
- Reverse PCR primer is complementary to the GTGG overhang at the 5' end: Make sure that the reverse PCR primer does not contain the sequence, CACC, at the 5' end.
- Use a thermostable, proofreading polymerase such as Accuprime Pfx DNA Polymerase (Cat. No. 12344024) to produce blunt-end PCR products.

What is the best molar ratio of PCR product:vector to use for TOPO TA cloning? Is there an equation to calculate the quantity to use?

We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio

I am cloning my gene of interest into the pENTR/SD/D-TOPO vector, but plan on expressing my gene in both E.coli and mammalian cells. Will the Shine-Dalgarno sequence interfere with mammalian expression?

No, the Shine-Dalgarno sequence does not adversely affect mammalian expression when used in an appropriate mammalian DEST vector.