PureLink™ PCR Purification Wash Buffer B1 is the wash buffer used with the PureLink™ PCR Purification Kit. The buffer isRead more
Have Questions?
Catalog Number
Quantity
K310006
40 mL
Catalog number K310006
Price (INR)
2,087.00
Online offer
Ends: 31-Mar-2026
3,478.00
Save 1,391.00 (40%)
Each
Quantity:
40 mL
Price (INR)
2,087.00
Online offer
Ends: 31-Mar-2026
3,478.00
Save 1,391.00 (40%)
Each
PureLink™ PCR Purification Wash Buffer B1 is the wash buffer used with the PureLink™ PCR Purification Kit. The buffer is activated by the addition of 160 mls of 100% ethanol.
For Research Use Only. Not for use in diagnostic procedures.
Can I use the PureLink PCR Purification Kit to do a gel purification?
The buffers and procedure are different for the PureLink PCR Purification Kit and PureLink Gel Purification Kit. Therefore, you can swap the column, but the right buffer and protocol must be used.
What are the minimum and maximum size ranges for DNA fragment cleanup using the PureLink PCR Clean-Up Kit?
The size range is between 100 bp and 12 kb. Our kit comes with two buffers, Binding Buffer (B2) and Binding Buffer HC (B3). The Binding Buffer HC (B3) eliminates primer-dimers and short failed PCR products that are smaller than 300 bp.