Product Image
DES® TOPO® TA Expression Kit *DISCONTINUED*
Have Questions?
Catalog number K412540
also known as K4125-40
Price (INR)
-
The DES® TOPO® TA Expression Kit offers one-step cloning of PCR products directly into an inducible DES® expression vector. The kit uses the linearized, topoisomerase I-activated pMT/V5-His-TOPO® vector for fast and easy cloning and subsequent high-level expression. Topoisomerase activation of this vector allows PCR products to be ligated in just 5 minutes on your bench top to produce >85%recombinants. The pMT/V5-His-TOPO® vector has all of the features of the pMT/V5-His vector to simplify protein expression, detection, and purification of recombinant proteins in Drosophila S2 cells.
Specifications
Unit Size40 reactions