TOPO&trade; TA Cloning&trade; Kit for Sequencing, with One Shot&trade; TOP10 Electrocomp&trade; <i>E. coli</i>
Invitrogen™

TOPO™ TA Cloning™ Kit for Sequencing, with One Shot™ TOP10 Electrocomp™ E. coli

The TOPO™ TA Cloning™ Kits for Sequencing provide a highly efficient, 5 minute, one-step cloning strategy ('TOPO™ Cloning') for theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K45800125 Reactions
K458040
also known as K4580-40
50 Reactions
Catalog number K458001
Price (INR)
-
Quantity:
25 Reactions
The TOPO™ TA Cloning™ Kits for Sequencing provide a highly efficient, 5 minute, one-step cloning strategy ('TOPO™ Cloning') for the direct insertion of Taq polymerase-amplified PCR products into a plasmid vector for sequencing. Each kit uses the pCR™4-TOPO™ TA vector with specially designed sequencing primer sites that return more insert sequence and less vector sequence from each reaction. These kits include everything necessary to clone and select recombinant vectors containing your PCR fragment of choice.

• Get More Sequence—Allows for more insert sequence and less vector seuquence when using standard sequencing primers
• Fast and Easy—Go from PCR-to-clones in just 3 steps and in as little as 5 minutes hands-on time
• Efficient—Achieve up to 95% clones with correct insert
• Proven—Reliable performance for over a decade with over 4000 citations

TOPO™ TA Cloning™ Kits for Sequencing Kit Overview
• VECTOR: pCR™4-TOPO™ TA Vector—Optimized cloning vector for improved sequencing results
• CLONING METHOD: TOPO™ TA Cloning —Topoisomerase I based 5 minute ligation of PCR products with 3´A overhangs (Taq-amplified) to the vector
• COMPETENT CELLS: Various Options —Choose from kits with either general, high-efficiency, bacteriophage T1-resistant, or fast-growing competent cells

pCR™4-TOPO™ TA Vector—Optimized for Sequencing
We have removed much of the multiple cloning site from the pCR™4-TOPO™ TA vector to shorten the distance between sequencing primer sites and the insert site to as little as 33 bp. This means sequencing reactions give less vector sequence and more insert sequence. The pCR™4-TOPO™ TA vector has sites for 4 common sequencing primers: M13 forward, M13 reverse, T7, and T3. The kits include an aliquot of each.

pCR™4-TOPO™ TA Clone Selection and Manipulation
The pCR™4-TOPO™ TA vector contains both ampicillin and kanamycin resistance markers and a LacZα-ccdB gene fusion for positive selection and blue/white screening. The vector’s minimized multiple cloning site still includes flanking EcoRI sites for simplified excision of cloned PCR products and a unique Sse8387I site for generation of nested deletions prior to sequencing. T7 and T3 promoters are also present for in vitro transcription.

Simplified TOPO™-Based Cloning
Using TOPO™ cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform with the providedE. coli competent cells.

Efficient Cloning
With up to 95% of clones carrying the desired insert, you can screen less clones to help you save time and money. The pCR™4-TOPO™ TA vector used in this kit comes with 3´T overhangs for efficient ligation of Taq-amplified PCR products with 3´A overhangs.

The Standard in Cloning
When it comes to cloning, TOPO™ cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO™ cloning has been applied to many different vectors for a wide array of applications.

TOPO™ TA Cloning™ Kits for Sequencing Kit Options
The TOPO™ TA Cloning™ Kit for Sequencing can be purchased with a variety of competent cells that deliver different advantages depending on what your needs are:

• General cloning: TOP10 (Cat. No. K4575-J10, K4575-01, K4575-40)
• High efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K4580-01, K4580-40)
• General cloning, bacteriophage T1 resistance: DH5α-T1R (Cat. No. K4595-01, K4595-40)
• Fast growth: Mach1™-T1R Chemically Competent E. Coli (Cat. No. K4530-20)

We also offer a version of the kit that includes a PureLink™ Quick Plasmid Miniprep Kit (Cat. No. K4575-02) for use in isolation of clean, sequencing-ready, recombinant plasmid.

TOPO™ products are For Research Use Only. Not intended for any animal or human therapeutic of diagnostic use.

Related Links
• Custom Vector Construction and Cloning Services
• Plasmid DNA Purification Kit Selection Guide
• PCR Reagents, Instruments, and Supplies
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10
Cloning MethodTOPO™-TA
For Use With (Application)Chromatin Biology
Product LineOne Shot
Product TypeCloning Kit
PromoterT7, T3
Quantity25 Reactions
VectorTOPO-TA Cloning Vectors
FormatKit
Unit SizeEach
Contents & Storage
Box 1:
• Topoisomerase I-activated pCR™4-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• T3, T7, M13 forward, and M13 reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.

Frequently asked questions (FAQs)

Can I store my competent E. coli in liquid nitrogen?

We do not recommend storing competent E. coli strains in liquid nitrogen as the extreme temperature can be harmful to the cells. Also, the plastic storage vials are not intended to withstand the extreme temperature and may crack or break.

How should I store my competent E. coli?

We recommend storing our competent E. coli strains at -80°C. Storage at warmer temperatures, even for a brief period of time, will significantly decrease transformation efficiency.

What is the best molar ratio of PCR product:vector to use for TOPO TA cloning? Is there an equation to calculate the quantity to use?

We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio

Does Platinum Taq DNA Polymerase High Fidelity enzyme mix leave 3' A-overhangs on the PCR product for subsequent cloning into a TOPO TA or original TA vector?

Yes, the enzyme mix leaves 3' A-overhangs on a portion of the PCR products. However, the cloning efficiency is greatly decreased compared to that obtained with Taq polymerase alone. It is recommended to add 3' A-overhangs to the product for TA cloning.