LysoSensor™ Yellow/Blue dextran, 10,000 MW, Anionic, Fixable
Invitrogen™

LysoSensor™ Yellow/Blue dextran, 10,000 MW, Anionic, Fixable

LysoSensor Yellow/Blue dextran is a ratiometric probe that can be used to measure the pH of acidic organelles, such asRead more
Have Questions?
Catalog NumberQuantity
L224605 mg
Catalog number L22460
Price (INR)
-
Quantity:
5 mg
LysoSensor Yellow/Blue dextran is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ∼3.9.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission Wavelength335/452
Quantity5 mg
Shipping ConditionRoom Temperature
Sub Cellular LocalizationLysosomes
ColorYellow, Blue
For Use With (Equipment)Fluorescence Microscope
Product LineLysoSensor
Product TypeDextran
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

Can I use a plate reader to measure the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes?

Using a plate reader for measuring the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes, provides weak pH measurements, as discrete regions of interest cannot be selected for analysis. Therefore, there is no way of excluding extracellular fluorescence and little to no spatial or morphological information can be obtained. Instead, a readout of whole-cell fluorescence is provided, resulting in the average intraorganellar pH of all labeled organelles for the analyzed cell population. The is also the risk that a signal may be dominated by a few extremely bright, but essentially irrelevant cells, such as dead cells, or dye-labeled debris particles.

For more information about measuring luminal pH in organelles, please see the reference linked below:

Live-cell Microscopy and Fluorescence-based Measurement of Luminal pH in Intracellular Organelles

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What reagents would you recommend for measuring the pH in acidic organelles, such as lysosomes?

For measuring the pH in acidic organelles, such as lysosomes, we recommend our LysoSensor ratio metric probes. Please see the links to the product page and descriptions below:

LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460):
LysoSensor Yellow/Blue dextran is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ~3.9.

LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545):
LysoSensor Yellow/Blue DND-160 is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue DND-160 is ~4.2.

LysoSensor Blue DND-167 - Special Packaging (Cat. No. L7533):
LysoSensor Blue DND-167 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Blue DND-167 is ~5.1.

LysoSensor Green DND-189 - Special Packaging (Cat. No. L7535):
LysoSensor Green DND-189 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Green DND-189 is ~5.2.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Could you provide me a protocol or reference for measuring the pH in acidic organelles, such as lysosomes?

Please find a link to a reference for measuring luminal pH in intracellular organelles below:

Live-cell Microscopy and Fluorescence-based Measurement of Luminal pH in Intracellular Organelles

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460) and LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545)?

LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460) has an emission of 335/452, is fixable and allows loading of cells by endocytosis. The pKa of LysoSensor Yellow/Blue dextran is ~3.9, which is slightly lower than LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545).

LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545) has an emission of 329/440 and is unique as it shows both dual-excitation and dual-emission spectral peaks that are pH-dependent. Nevertheless, this LysoSensor only exhibits the pH-dependent dual-emission spectra in living cells. Lysosensor Yellow/Blue DND-160 has predominantly yellow fluorescence, and in less acidic organelles is has blue fluorescence. Dual-emission measurements may permit ratio imaging of the pH in acidic organelles such as lysosome.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (15)

Citations & References
Abstract
Influence of P-glycoprotein inhibitors on accumulation of macrolides in J774 murine macrophages.
Authors:Seral C, Michot JM, Chanteux H, Mingeot-Leclercq MP, Tulkens PM, Van Bambeke F
Journal:Antimicrob Agents Chemother
PubMed ID:12604540
'The influence of inhibitors of P-glycoprotein (verapamil [VE], cyclosporine [CY], and GF120918 [GF]) on the cell handling of macrolides (erythromycin [ERY], clarithromycin [CLR], roxithromycin [ROX], azithromycin [AZM], and telithromycin [TEL]) was examined in J774 murine macrophages. The net influx rates of AZM and TEL were increased from 2- to 3.5-fold ... More
d-Tocopherol Reduces Lipid Accumulation in Niemann-Pick Type C1 and Wolman Cholesterol Storage Disorders.
Authors:Xu M, Liu K, Swaroop M, Porter FD, Sidhu R, Finkes S, Ory DS, Marugan JJ, Xiao J, Southall N, Pavan WJ, Davidson C, Walkley SU, Remaley AT, Baxa U, Sun W, McKew JC, Austin CP, Zheng W,
Journal:J Biol Chem
PubMed ID:23035117
'Niemann-Pick disease type C (NPC) and Wolman disease are two members of a family of storage disorders caused by mutations of genes encoding lysosomal proteins. Deficiency in function of either the NPC1 or NPC2 protein in NPC disease or lysosomal acid lipase in Wolman disease results in defective cellular cholesterol ... More
Lysosomal proteolysis and autophagy require presenilin 1 and are disrupted by Alzheimer-related PS1 mutations.
Authors:Lee JH, Yu WH, Kumar A, Lee S, Mohan PS, Peterhoff CM, Wolfe DM, Martinez-Vicente M, Massey AC, Sovak G, Uchiyama Y, Westaway D, Cuervo AM, Nixon RA,
Journal:Cell
PubMed ID:20541250
Macroautophagy is a lysosomal degradative pathway essential for neuron survival. Here, we show that macroautophagy requires the Alzheimer's disease (AD)-related protein presenilin-1 (PS1). In PS1 null blastocysts, neurons from mice hypomorphic for PS1 or conditionally depleted of PS1, substrate proteolysis and autophagosome clearance during macroautophagy are prevented as a result ... More
Cytokines regulate proteolysis in major histocompatibility complex class II-dependent antigen presentation by dendritic cells.
Authors:Fiebiger E, Meraner P, Weber E, Fang IF, Stingl G, Ploegh H, Maurer D
Journal:J Exp Med
PubMed ID:11304549
Endo/lysosomal proteases control two key events in antigen (Ag) presentation: the degradation of protein Ag and the generation of peptide-receptive major histocompatibility complex (MHC) class II molecules. Here we show that the proinflammatory cytokines tumor necrosis factor alpha and interleukin (IL)-1beta rapidly increase the activity of cathepsin (cat) S and ... More
Optimal transfection with the HK polymer depends on its degree of branching and the pH of endocytic vesicles.
Authors:Chen QR, Zhang L, Luther PW, Mixson AJ
Journal:Nucleic Acids Res
PubMed ID:11884631
We have recently reported that liposomes in combination with histidine (HK)-containing polymers enhanced the expression of luciferase in transfected cells. In transformed or malignant cell lines, branched HK polymers (combined with liposome carriers) were significantly more effective than the linear HK polymer in stimulating gene expression. In the current study, ... More