Lipopolysaccharides From <i>Escherichia coli</i> Serotype 055:B5, Alexa Fluor&trade; 568 Conjugate
Invitrogen™

Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 568 Conjugate

This bright, orange-red—fluorescent Alexa Fluor 568 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.Read more
Have Questions?
Catalog NumberQuantity
L23352100 μg
Catalog number L23352
Price (INR)
18,615.00
キャンペーン価格
Ends: 30-Sep-2026
26,592.00
Save 7,977.00 (30%)
Each
Quantity:
100 μg
Price (INR)
18,615.00
キャンペーン価格
Ends: 30-Sep-2026
26,592.00
Save 7,977.00 (30%)
Each
This bright, orange-red—fluorescent Alexa Fluor 568 conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAlexa Fluor Dyes
Excitation/Emission578/603
FormSolid
Quantity100 μg
Shipping ConditionRoom Temperature
Product LineAlexa Fluor
Product TypeLipopolysaccharide Conjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (3)

Citations & References
Abstract
A Single Human VH-gene Allows for a Broad-Spectrum Antibody Response Targeting Bacterial Lipopolysaccharides in the Blood.
Authors:
Journal:Cell Rep
PubMed ID:32846123
Taxifolin ameliorates sepsis-induced lung capillary leak through inhibiting the JAK/STAT3 pathway.
Authors:
Journal:Allergol Immunopathol (Madr)
PubMed ID:35257540
Modeling hypercholesterolemia and vascular lipid accumulation in LDL receptor mutant zebrafish.
Authors:
Journal:J Lipid Res
PubMed ID:29187523