Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL)
Invitrogen™

Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL)

Human low-density lipoprotein (LDL) is large protein complex (∼500,000 Da) that binds to a specific receptor on the surface ofRead more
Have Questions?
Catalog NumberQuantity
L35354200 μL
Catalog number L35354
Price (INR)
21,588.00
Online offer
Ends: 31-Dec-2026
31,982.00
Save 10,394.00 (32%)
Each
Quantity:
200 μL
Price (INR)
21,588.00
Online offer
Ends: 31-Dec-2026
31,982.00
Save 10,394.00 (32%)
Each
Human low-density lipoprotein (LDL) is large protein complex (∼500,000 Da) that binds to a specific receptor on the surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis—our labeled LDL complexes are useful tools for studying this phenomenon. These experiments are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of the label can be analyzed after the specified time period. We offer an unlabeled LDL and two classes of labeled LDLs: those containing an unmodified apoprotein (used to study normal cholesterol delivery and internalization) and those with an acetylated (Ac) apoprotein (used to study cell types that express receptors specific for this acetylated version (i.e., endothelial and microglial cells)).

LDL Specifications:

• Label (Ex/Em): Unlabeled
• Acetylated: Yes
• Amount: 200 μL (2.5 mg/mL)

Fresh LDL Produced Continually
We prepare our LDL and AcLDL products from fresh human plasma approximately every two months. The nonacetylated LDL products are shipped within two weeks of their preparation. All acetylated LDL products are available on a continuous basis.

Nonacetylated vs. Acetylated LDL
LDL containing an unmodified apoprotein is used to study normal cholesterol delivery and internalization. If the lysine residues of LDL’s apoprotein have been acetylated, the LDL complex no longer binds to the LDL receptor, but rather is taken up by endothelial and microglial cells that possess “scavenger” receptors specific for that modified form.

Key Applications for Labeled LDL
Some of the many applications for labeled LDL complexes include:
• Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization
• Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay)
• Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration2.5 mg⁄ml
Detection MethodFluorescence
Dye TypeUnlabeled
FormLiquid
Quantity200 μL
Shipping ConditionWet Ice
Product TypeSupplement
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?

We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use Low Density Lipoprotein (LDL) from Human Plasma on other species?

We have not tested this.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (17)

Citations & References
Abstract
Activation of proteinase-activated receptor-2 in mesothelial cells induces pleural inflammation.
Authors:Lee YC, Knight DA, Lane KB, Cheng DS, Koay MA, Teixeira LR, Nesbitt JC, Chambers RC, Thompson PJ, Light RW
Journal:Am J Physiol Lung Cell Mol Physiol
PubMed ID:15591415
'Pleural inflammation underlies many pleural diseases, but its pathogenesis remains unclear. Proteinase-activated receptor-2 (PAR(2)) is a novel seven-transmembrane receptor with immunoregulatory roles. We hypothesized that PAR(2) is present on mesothelial cells and can induce pleural inflammation. PAR(2) was detected by immunohistochemistry in all (19 parietal and 11 visceral) human pleural ... More
N-glycans of SREC-I (scavenger receptor expressed by endothelial cells): essential role for ligand binding, trafficking and stability.
Authors:Sano M, Korekane H, Ohtsubo K, Yamaguchi Y, Kato M, Shibukawa Y, Tajiri M, Adachi H, Wada Y, Asahi M, Taniguchi N,
Journal:Glycobiology
PubMed ID:22279061
'Scavenger receptor expressed by endothelial cells (SREC-I) mediates the endocytosis of chemically modified lipoproteins such as acetylated low-density lipoprotein (Ac-LDL) and oxidized LDL and is implicated in atherogenesis. We produced recombinant SREC-I in Chinese hamster ovary-K1 cells and identified three potential glycosylation sites, Asn(289), Asn(382) and Asn(393), which were all ... More
Uptake and trafficking of liposomes to the endoplasmic reticulum.
Authors:Pollock S, Antrobus R, Newton L, Kampa B, Rossa J, Latham S, Nichita NB, Dwek RA, Zitzmann N,
Journal:FASEB J
PubMed ID:20097877
'Liposomes are vesicular structures consisting of an aqueous core surrounded by a lipid bilayer. Apart from the cytosol and lysosomes, no other intracellular compartment has been successfully targeted using liposomal delivery. Here, we report the development of liposomes capable of specific targeting to the endoplasmic reticulum (ER) and associated membranes. ... More
Scavenger receptor classes A and B. Their roles in atherogenesis and the metabolism of modified LDL and HDL.
Authors:Van Berkel TJ, Van Eck M, Herijgers N, Fluiter K, Nion S
Journal:Ann N Y Acad Sci
PubMed ID:10865831
'Scavenger-receptor class A has been held responsible for the clearance of modified LDL from the blood circulation. However, in mice deficient in scavenger-receptor class A, the decay in vivo of acetylated LDL (t1/2 < 2 min), as well as tissue distribution and liver uptake (at 5 min 77.4 +/- 4.6% ... More
Role of phospholipid transfer protein in high-density lipoprotein metabolism: insights from studies in transgenic mice.
Authors:Van Tol A, Jauhiainen M, De Crom R, Ehnholm C
Journal:Int J Tissue React
PubMed ID:10937357