NIR Standards and References
Actual product may vary
Thermo Scientific™
NIR Standards and References
Help you fulfill regulatory requirements using certified and traceable standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
LABSPHEREReflection Linearity Standard
KTA-1920XWavelength Accuracy Standard
NICOLETWAVELENGTHStandard
NICOLETSTANDARDSKTLinearity Standard
Catalog number LABSPHERE
Price (INR)
-
Type:
Reflection Linearity Standard
Thermo Scientific™ NIR Standards and References for Antaris FT-NIR help you fulfill regulatory requirements using certified and traceable standards. These industry-recommended standards can be incorporated into the ValPro™ Qualification testing routines. They also supplement the NIST-traceable and serialized validation wheel available with the ValPro System Qualification Package.
Standard reference materials are available for both reflectance and transmission sampling techniques.
External Reflectance Standards
  • Reflection Standards Kit consisting of six linearity standards-99%, 80%, 40%, 20%, 10% and 2% reflective
  • KTA-1920x NIST™-Traceable Reflection Standard for the measurement of wavelength accuracy and precision
  • 99% Reflective Diffuse Gold Reflection Standard
  • Polystyrene Wavelength reference sample
  • 99% Reflective Spectralon reference sample
External Transmission Standards
  • Transmission Standards Kit consisting of five linearity standards-80%, 40%, 20%, 10% and 2% transmission
  • NIST-Traceable Polystyrene Wavelength Transmission standard
Specifications
For Use With (Equipment)Antaris FT-NIR
TypeReflection Linearity Standard
Unit SizeEach