Barnstead™ Nanopure™ Accessories and Consumables
Actual product may vary
Thermo Scientific™
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
LMX26UV Lamp (TOC)
D50280R/O and Distilled Feed (Ultralow Organics, Type I Water)
D50281Deionized Water Feed (Ultralow Organics, Type I Water)
D50283Deionized Water Feed (Low Organics, Type I Water)
D8964Low Water Level Pump Protector
E896X5Calibration Module
Catalog number LMX26
Price (INR)
-
Type:
UV Lamp (TOC)
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
For Use With (Equipment)Nanopure Water Purifications System
TypeUV Lamp (TOC)
Unit SizeEach