Oxoid™ Gelatin Bacteriological
Thermo Scientific™
Oxoid™ Gelatin Bacteriological
Oxoid Gelatin Bacteriological is a special bacteriological grade readily soluble in water to give a clear solution free from sulphite and preservatives.
Have Questions?
Catalog NumberDescription
LP0008BDehydrated; Gelatin bacteriological
Catalog number LP0008B
Price (INR)
-
Description:
Dehydrated; Gelatin bacteriological
Oxoid Gelatin Bacteriological is a special bacteriological grade readily soluble in water to give a clear solution free from sulphite and preservatives.
  • Firm gel obtained from 15% solution
Specifications
DescriptionDehydrated; Gelatin bacteriological
Quantity500 g
FormPowder
Product TypeGelatin
Unit SizeEach