Oxoid Agar Technical (Agar No. 2) is a general-purpose bacteriological-grade agar.
Have Questions?
Change view
Catalog Number
Quantity
LP0012B
500 g
LP0012T
5 kg
2 Options
Catalog number LP0012B
Price (INR)
-
Quantity:
500 g
A technical grade agar which conforms to the specifications of the U.S. Pharmacopoeia and A.P.H.A. Suitable for applications where clarity and compatibility are not of prime importance. It is of high gel strength and will give a firm gel 1.2% w/v.