Oxoid™ Agar Technical No.2
Thermo Scientific™
Oxoid™ Agar Technical No.2
Oxoid Agar Technical (Agar No. 2) is a general-purpose bacteriological-grade agar.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0012T5 kg
LP0012B500 g
Catalog number LP0012T
Price (INR)
-
Quantity:
5 kg
A technical grade agar which conforms to the specifications of the U.S. Pharmacopoeia and A.P.H.A. Suitable for applications where clarity and compatibility are not of prime importance. It is of high gel strength and will give a firm gel 1.2% w/v.
Specifications
Quantity5 kg
Product TypeAgar
Unit SizeEach