Oxoid™ Liver Digest Neutralized
Thermo Scientific™

Oxoid™ Liver Digest Neutralized

The Oxoid Liver Digest Neutralized is a biologically standardized papaic digest of ox liver.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0027B500 g
LP0027T5 kg
LP0027K25 kg
Catalog number LP0027B
Price (INR)
-
Quantity:
500 g

Thermo Scientific™ Oxoid Liver Digest Neutralized is a biologically standardized papaic digest of ox liver for use as a source of nutrients in microbiological culture media or fermentation.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Culture media or fermentation processes
Protein SourceBeef liver
Quantity500 g
Shelf Life720 Days from Manufacture
YieldFor 2.5L medium
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage