Oxoid™ Peptonised Milk
Thermo Scientific™
Oxoid™ Peptonised Milk
Oxoid Peptonised Milk is a pancreatic digest of high grade skimmed milk powder with high levels of carbohydrate and calcium.
Have Questions?
Catalog NumberQuantity
LP0032J250g
Catalog number LP0032J
Price (INR)
-
Quantity:
250g
Oxoid Peptonised Milk is a pancreatic digest of high grade skimmed milk powder with high levels of carbohydrate and calcium.
  • Particularly useful in media for bacteriological examination of dairy products
Specifications
DescriptionPeptonized milk
Quantity250g
FormPowder
Product TypeMilk
Unit SizeEach