Oxoid™ Tryptone T
Actual product may vary
Thermo Scientific™

Oxoid™ Tryptone T

Oxoid Tryptone T peptone is optimized for low ion levels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0043T5 kg
LP0043C400 g
LP0043B500 g
Catalog number LP0043T
Price (INR)
-
Quantity:
5 kg
Thermo Scientific™ Oxoid Tryptone T peptone developed with lower ion levels is ideal for the production of toxin by Clostridium tetani in fermentation applications.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Fermentation processes
Protein SourceCasein
Quantity5 kg
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage