Oxoid™ Proteose Peptone
Thermo Scientific™

Oxoid™ Proteose Peptone

Oxoid Proteose Peptone is a specialty peptone containing proteoses as defined in the USP.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0085B500 g
LP0085T5 kg
Catalog number LP0085B
Price (INR)
-
Quantity:
500 g
Thermo Scientific™ Oxoid Proteose Peptone is a specialty peptone containing proteoses as defined in the USP.

Particularly suitable for use in toxin production and fermentation applications or in culture media for bacteria with wide-ranging nutritional requirements.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Certifications/ComplianceIndustrial Reference: USP.
For Use With (Application)Culture media or fermentation processes
Protein SourcePork / Beef / Soy
Quantity500 g
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage