Bacteriological Peptone No. 2
Thermo Scientific™

Bacteriological Peptone No. 2

The Oxoid Peptone Bacteriological is an all-purpose peptone specially prepared for use in fermentation applications - exclusively from New Zealand sourced beef.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0137K25 kg
LP0137B500 g
Catalog number LP0137K
Price (INR)
-
Quantity:
25 kg
Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralised form are very nutritious all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. They are specially prepared to be compatible with other refined culture media or fermentation ingredients.

The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles. 

The neutralised form evolved from the original to meet those occasions when a slightly higher pH is required. Both when reconstituted give a solution free of haze, cloudiness or precipitation.

LP0137 is manufactured using beef from New Zealand.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Culture media or fermentation processes
Media TypeBacteriological Peptone No. 2
Protein SourceBeef
Quantity25 kg
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Manufacturing OriginUnited Kingdom
Product TypeMedia Additive
SterilityNon-sterile
Target Organism ClassBacteria
Test TypeMicrobiological Tests
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage