Oxoid™ Peptone P
Oxoid™ Peptone P
Thermo Scientific™

Oxoid™ Peptone P

Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
LP0149G10 kg
LP0149B500 g
LP0149T5 kg
Catalog number LP0149G
Price (INR)
-
Quantity:
10 kg
Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification of peptic digests of animal tissue.

The high sulphur content makes it ideal for use in culture media.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
For Use With (Application)Culture media
Protein SourcePork
Quantity10 kg
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage