RevertAid™ Master Mix
Thermo Scientific™

RevertAid™ Master Mix

Thermo Scientific RevertAid Master Mix is a complete system for efficient synthesis of first-strand cDNA from mRNA or total RNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
M163150 Reactions
M1632100 Reactions
Catalog number M1631
Price (INR)
-
No. of Reactions:
50 Reactions

Thermo Scientific RevertAid Master Mix is a complete system for efficient synthesis of first-strand cDNA from mRNA or total RNA templates. The mix includes RevertAid Reverse Transcriptase (RT), which has lower RNase H activity compared to AMV reverse transcriptase. RevertAid Master Mix provides all cDNA synthesis reaction components in a convenient two-tube master mix. It is optimized for highly efficient cDNA synthesis for two-step quantitative RT-PCR (RT-qPCR) applications. The two-tube formulation has a shorter reaction time and requires fewer pipetting steps than the RevertAid RT Reverse Transcription Kit, while maintaining high performance.

Highlights
• Fast—short reaction time (typically 15–30 min)
• Convenient—easy workflow with few pipetting steps
• Reliable—reverse transcription of both abundant and limited targets
• Flexible—optimized 2-step protocol, enabling multiple PCR reactions from a single reverse-transcription reaction

Applications
First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatMaster Mix
No. of Reactions50 Reactions
Quantity50 reactions
Reaction FormatPremixed Components
Reagent TypeReverse Transcription
Reverse TranscriptaseRevertAid
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)Real Time PCR (qPCR)
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• 20X Enzyme mix (1 x 50 μL)
• 2X RT Buffer Mix (1 x 500 μL)

Store at –15 to –25°C.
2X RT Buffer Mix can be stored at 2–8°C for up to 6 months.