Microplates for Fluorescence-based Assays
Invitrogen™

Microplates for Fluorescence-based Assays

Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsQuantity
M3309138410 Plates
M330899610 Plates
M330909630 Plates
Catalog number M33091
Price (INR)
14,810.00
線上優惠
Ends: 30-Sep-2026
21,156.00
Save 6,346.00 (30%)
10 microplates
No. of Wells:
384
Quantity:
10 Plates
Price (INR)
14,810.00
線上優惠
Ends: 30-Sep-2026
21,156.00
Save 6,346.00 (30%)
10 microplates

Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).

Features of the microplates for fluorescence-based assays include:

  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AutoclavableNot Autoclavable
DisposableSingle-use
For Use With (Application)Nucleic Acid Quantification
FormatStandard
Packaging10-plates
Product TypeMicroplates
Quantity10 Plates
Volume (Metric) Working40 μL
Array16 x 24
BarcodeNo Barcode
ColorClear Bottom, Black
MaterialPolystyrene
No. of Wells384
Product LineInvitrogen
SterilityNon-sterile
Well ShapeFlat Bottom (F-well)
Unit Size10 microplates
Contents & Storage
Store at room temperature.