Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
Actual product may vary
Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™
Fisher Chemicals

Metaphosphoric Acid, Extra Pure, SLR, Fisher Chemical™

HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979 | CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Catalog NumberQuantityPackaging
M385153
also known as M/3851/53
500 gPlastic powder jar
Catalog number M385153
also known as M/3851/53
HSN 28092020
Price (INR)
30,779.00
Each
Quantity:
500 g
Packaging:
Plastic powder jar
Price (INR)
30,779.00
Each
HO3P, CAS Number-37267-86-0, unii-mtk99r3uv0, x-phosphate-group, m-phosphoric acid, metaphosphoric acid hpo3, meta-phosphoric acid, phosphenic acid, metaphosphoric acid, phosphonate 1-, hydrogenphosphonate, hydrogen phosphonate, 500g, Colorless, UEZVMMHDMIWARA-UHFFFAOYSA-N, 200 deg.C, 11351, 79.979
Specifications
pH2
ColorColorless
CAS37267-86-0
Melting Point200°C
PackagingPlastic powder jar
Quantity500 g
Molecular FormulaHO3P
Physical FormSolid
MDL Number11351
Synonymmetaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
InChI KeyUEZVMMHDMIWARA-UHFFFAOYSA-N
SMILESOP(=O)=O
Molecular Weight (g/mol)79.979
PubChem CID3084658
Unit SizeEach