Pathatrix™ Magnetic Capture Plate
Applied Biosystems™

Pathatrix™ Magnetic Capture Plate

The Pathatrix™ Magnetic Capture Plate makes it easy to link to endpoint detection by real-time PCR using the DuPont BAX™ system.
Have Questions?
Catalog NumberQuantity
MAGNETICPLATE1 Each
Catalog number MAGNETICPLATE
Price (INR)
-
Quantity:
1 Each

The Pathatrix™ Magnetic Capture Plate makes it easy to link to endpoint detection by real-time PCR using the DuPont BAX™ system. The magnetic rack is used to remove the paramagnetic particles after lysis and prior to detection.

Specifications
Capacity (Metric)1 x 96-well Plate
For Use With (Equipment)Pathatrix™ Auto System
Format96-well plate
Quantity1 Each
Shipping ConditionRoom Temperature
Product LinePathatrix
TypeMagnetic Capture Plate
Unit SizeEach
Contents & Storage
1 plate

Store at room temperature.