Optical Filter for Multiskan™ Ease
Optical Filter for Multiskan™ Ease
Optical Filter for Multiskan™ Ease
Thermo Scientific™
Optical Filter for Multiskan™ Ease
The Thermo Scientific™ filter accessories for the Multiskan™ Ease Absorbance Microplate Reader expand assay flexibility by enabling precise wavelength selection across a wide range of absorbance applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWavelength
MEOF0690690 nm
MEOF0414414 nm
MEOF0492492 nm
MEOF0520520 nm
MEOF0540540 nm
MEOF0570570 nm
MEOF0600600 nm
MEOF0660660 nm
MEOF0750750nm
Catalog number MEOF0690
Price (INR)
48,207.00
Each
Wavelength:
690 nm
Price (INR)
48,207.00
Each
The Thermo Scientific™ filter accessories for the Multiskan™ Ease Absorbance Microplate Reader expand assay flexibility by enabling precise wavelength selection across a wide range of absorbance applications. Designed to integrate smoothly with the instrument's optical system, these durable hard-coated filters support both routine and advanced assays while maintaining high measurement accuracy and long-term reliability.
The Multiskan™ Ease utilizes an easy-to-install 8-position filter wheel, supplied with three pre-installed standard filters (405 nm, 450 nm, and 620 nm). Additional filter accessories allow users to customize wavelength configurations to meet specific assay requirements, from ELISA and protein quantification to enzyme kinetics and microbial growth analysis.
Optical Filter Wavelengths and Typical Applications
Standard (pre-installed on filter wheel):
  • 405 nm – Alkaline phosphatase (ALP) ELISA with pNPP substrate; ABTS assays; enzyme kinetics; endotoxin (LAL) assays
  • 450 nm – HRP/TMB-based ELISA (including Rapid and Instant ELISA formats)
  • 620 nm – ELISA reference wavelength; alternative read for Bradford protein assays
Optional filters:
  • 414 nm – Heme/hemoglobin assays, Soret band detection, selected oxidative and antioxidant capacity assays
  • 492 nm – OPD-based ELISA, LDH cytotoxicity, MTS cell proliferation assays. Pierce™ Dilution-Free™ Rapid Gold BCA Protein Assay.
  • 520 nm – Selected colorimetric cell proliferation and metabolic activity assays
  • 540 nm – Nitric oxide (Griess) assays
  • 570 nm – Primary wavelength for BCA protein quantification, MTT cell proliferation and crystal violet assays
  • 600 nm – Bacterial growth (OD600), Bradford protein assay and turbidity measurements
  • 660 nm – Pierce™ 660 nm protein assay
  • 690 nm – Reference wavelength for ELISA and MTT assays
  • 750 nm – Lowry protein assays; reference wavelength for protein assays and turbidity correction
Key Features
  • Easy-to-install design for quick filter changes without tools
  • Durable hard-coated optical filters designed for long lifetime performance and minimal maintenance
  • Optimized wavelength accuracy for reliable, reproducible absorbance measurements
  • Compatible with the 8-position filter wheel of the Multiskan™ Ease
These optical filters and filter wheel are designed for use with the Thermo Scientific™ Multiskan™ Ease Absorbance Microplate Reader. They support assay-specific wavelength optimization to enhance data quality and are fully integrated with Thermo Scientific™ SkanIt™ Software for streamlined assay setup and data analysis within existing laboratory workflows.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithReference wavelength for ELISA and MTT assays
Quantity1 Ea.
Wavelength690 nm
For Use With (Equipment)Multiskan Ease
Product TypeOptical Filter
Unit SizeEach