Dial Thermometer for Explosion-Proof Hotplates
Dial Thermometer for Explosion-Proof Hotplates
Thermo Scientific™
Dial Thermometer for Explosion-Proof Hotplates
Monitor top-plate temperature on an explosive hotplate with the Thermo Scientific™ Dial Thermometer for Explosion-Proof Hotplates.
Have Questions?
Catalog NumberFor Use With (Equipment)
MEX126Explosion-Proof Hotplates
Catalog number MEX126
Price (INR)
-
Request A Quote
For Use With (Equipment):
Explosion-Proof Hotplates
Monitor top-plate temperature on an explosive hotplate with the Thermo Scientific™ Dial Thermometer for Explosion-Proof Hotplates.
  • Temperature range: 0° to 300°C
  • Stem Dia. × L: 0.64 x 6.4cm (0.25 x 2.5 in.)
  • Face Dia.: 2.5cm (1 in.)
Specifications
For Use With (Equipment)Explosion-Proof Hotplates
Temperature (Metric)0°C to 300°C
TypeThermometer
Unit SizeEach