InSight Mote Transmitters
InSight Mote Transmitters
Actual product may vary
Thermo Scientific™
InSight Mote Transmitters
Thermo Scientific™ InSight mote transmitters periodically wake up and transmit sensor data to the InSight link server.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChannels
MOTET3C1D2A111137
MOTET3C1D2A111108
Catalog number MOTET3C1D2A11113
Price (INR)
-
Request A Quote
Channels:
7
Thermo Scientific™ InSight battery operated motes are configured during installation for your wireless network. Periodically, they wake up and transmit sensor data to the InSight link server. In the case of communication failure, the mote will log data for up to 24 hours.
  • Motes are available for WiFi and 915MHz wireless servers (North America)
  • Warranty varies by country, please contact us for details
Not available outside North America
Specifications
Battery TypeC
CalibrationNot Calibrated
Channels7
DescriptionInSight 802.11(b,g,n) Wi-Fi sensor transmitter standard range, 7 channels: 3-temp, 1-KVA/50 amps, ext power connector, 2-door inputs, internal ambient temp, “C” battery and enclosure. External sensors not included.
Display TypeNo Display
TypeInsight Mote Transmitter
Unit SizeEach