Search
Search
| Catalog Number | Product Type |
|---|---|
| N03078 | Universal plate tray for plates without lid |
| VLUPL0002 | Luminometric Upgrade Kit |
| F42550 | Emission Filter |
| N02339 | Robotic plate tray adapter for plates without lid |
| N02690 | 384-well Adapter for Plate w/o Lid |
| N02691 | 384-well Adapter for Plate w/ Lid |
| N02692 | 96-well Adapter for Plate w/o Lid |
| N02697 | 6-48-well Adapter for Plate w/ Lid |
| N03395 | 96-well adapter for PCR plate without lid |
| SP-00093 | Dispenser syringe, 5 mL |
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.