Multidrop Combi nL Reagent Dispensing valve
Thermo Scientific™
Multidrop Combi nL Reagent Dispensing valve
Optimize dispensing results with this range of accessories.
Have Questions?
Catalog NumberQuantity
N07494Each
Catalog number N07494
Price (INR)
-
Quantity:
Each
Optimize dispensing results with Thermo Scientific™ Multidrop Combi nL Reagent Dispenser Acccessories.
Specifications
DescriptionDispensing valve
QuantityEach
Product TypeReagent Dispensing Valve
Unit SizeEach
Frequently asked questions (FAQs)
I have an old spare valve which does not have any valve correction factor enclosed. Can I still use it?
Yes, old spare valves do not have this correction factor and the default value 0 is correct. If an old valve replaces the newer version, the correction factor needs to be set to value 0.
Do I need to recalibrate the instrument when a dispensing valve is changed?
No, you do not have to recalibrate the instrument, because the valve performance is so consistent. However you must apply the valve correction factor to the instrument through the FillIt software.
What is the orifice diameter (OD) of the dispensing valve?
The orifice (OD) is 100 μm.