Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes
Invitrogen™

Neon™ NxT Electroporation System 10 μL Kit with 8-Channel Tubes

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
N1096-896 x 2 Reactions
N10384-8384 x 2 Reactions
Catalog number N1096-8
Price (INR)
144,431.00
온라인 행사
Ends: 31-Mar-2026
240,718.00
Save 96,287.00 (40%)
Each
No. of Reactions:
96 x 2 Reactions
Price (INR)
144,431.00
온라인 행사
Ends: 31-Mar-2026
240,718.00
Save 96,287.00 (40%)
Each

The Neon NxT Electroporation System 10-μL Kit with 8-Channel Tubes is designed specifically for use with the Neon NxT Electroporation System with 8-channel pipette and pipette station (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Use this kit for transfection volumes of 10 μL, containing 5 x 104 to 2 x 105 adherent cells or 1 x 105 to 5 x 105 suspension cells. Cells that have been transfected using the included 10-μL Neon NxT tips are ready to be plated into a multi-well culture dish.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineStem Cell, Primary Cell, Hard-to-Transfect Cell
For Use With (Application)Transfection
For Use With (Equipment)Neon™ NxT Electroporation System
No. of Reactions96 x 2 Reactions
Quantity96 x 2 Reactions
Volume (Metric) Sample10 μL
FormatElectroporation Pipette Tip
Transfection TechniqueElectroporation
Unit SizeEach
Contents & Storage
Box 1 includes:
• Resuspension R Buffer, 4 x 1 mL
• Resuspension T Buffer, 4 x 1 mL
• Resuspension Genome Editing Buffer, 4 x 1 mL
• Electrolytic E10 Buffer, 2 x 100 mL

Store at 4°C.

Box 2 includes:
• Reaction tips, 96 tips
• Electroporation 8-channel tubes, 4 tubes

Store at room temperature.