Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers

Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers

Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
N12785Storage Bottle CapWellwash Microplate Strip Washer
N12407Liquid FilterWellwash Microplate Strip Washer
Catalog number N12785
Price (INR)
4,386.00
Each
Type:
Storage Bottle Cap
For Use With (Equipment):
Wellwash Microplate Strip Washer
Price (INR)
4,386.00
Each
These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.
Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
Specifications
Quantity2 Pk.
For Use With (Equipment)Wellwash Microplate Strip Washer
TypeStorage Bottle Cap
Unit SizeEach