MicroAmp™ 12-Cap Strip
Applied Biosystems™

MicroAmp™ 12-Cap Strip

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
N8010534200 Strips
N80115341000 Strips (5 pk./200)
Catalog number N8010534
Price (INR)
7,920.00
Online offer
Ends: 31-Mar-2026
13,200.00
Save 5,280.00 (40%)
Each
Quantity:
200 Strips
Price (INR)
7,920.00
Online offer
Ends: 31-Mar-2026
13,200.00
Save 5,280.00 (40%)
Each

MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. Attach caps using the MicroAmp™ Cap Installing Tool.

Will not interfere with sample reading.

  • Provides tight seal to minimize evaporation
  • Easy to apply and remove

Minimizes Evaporation

Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.

Easy to Use

Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.

Colored for Easy Organization

MicroAmp™ Caps Strips are available in different colors to help you keep track of your tubes during assay preparation.

Note: See user's manual or package insert for limited label license and trademark information.

Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers
Format200 Strip Caps
No. of Wells96
Product LineMicroAmp™
Quantity200 Strips
Reaction SpeedStandard
Shipping ConditionRoom Temperature
Closure TypeDomed Cap
ColorClear
MaterialPolypropylene
Product Type12-cap Strip
Unit SizeEach
Contents & Storage
Includes 200 MicroAmp™ 12-Cap Strips, Clear. Store at room temperature.