MicroAmp™ 12-Cap Strip, assorted colors
Applied Biosystems™

MicroAmp™ 12-Cap Strip, assorted colors

MicroAmp® Cap Strips are designed to fit on MicroAmp® reaction tubes, tube strips, and 96-well plates. They are available inRead more
Have Questions?
Catalog NumberQuantity
N8010834200 Strips
Catalog number N8010834
Price (INR)
8,735.00
Online offer
Ends: 31-Mar-2026
14,558.00
Save 5,823.00 (40%)
Each
Quantity:
200 Strips
Price (INR)
8,735.00
Online offer
Ends: 31-Mar-2026
14,558.00
Save 5,823.00 (40%)
Each
MicroAmp® Cap Strips are designed to fit on MicroAmp® reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. Attach caps using the MicroAmp® Cap Installing Tool.

Will not interfere with sample reading

• Provides tight seal to minimize evaporation
• Easy to apply and remove

Minimizes Evaporation
Use MicroAmp® Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.

Easy to Use
Use the MicroAmp® Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.

Colored for Easy Organization
MicroAmp® Caps Strips are available in different colors to help you keep track of your tubes during assay preparation.

Note: See user's manual or package insert for limited label license and trademark information.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers
Format200 Strip Caps
No. of Wells96
Product LineMicroAmp™ 
Quantity200 Strips
Shipping ConditionRoom Temperature
Closure TypeDomed Cap
ColorAssorted
MaterialPolypropylene
Product Type12-Strip Cap
Volume (Metric)0.2 mL
Unit SizeEach
Contents & Storage
Contains 200 MicroAmp™ 12-Cap Strips, Assorted Colors. Store at room temperature.