Nanobuilder
Thermo Scientific™
Nanobuilder
Thermo Scientific™ NanoBuilder™ is a tool for systematic planning of construction of multi-layer nanostructures by dividing CAD files into ordered projects.
Have Questions?
Catalog number NANOBUILDER
Price (INR)
-
Thermo Scientific™ NanoBuilder™ is a tool for systematic planning of construction of multi-layer nanostructures by dividing CAD files into ordered projects.
Thermo Scientific™ NanoBuilder™ is a tool for systematic planning of construction of multi-layer nanostructures by dividing CAD files into ordered projects. This facilitates the generation of complex structures that previously were tedious or even impossible to build.NanoBuilder allows accurate patterning of large and complex nanostructures on multiple sites, supporting all DualBeam patterning processes: focused ion beam (FIB) milling, gas-assisted FIB milling, FIB induced deposition, and electron-induced deposition.NanoBuilder is installed on the control PC of the SDB tool. The package includes one NanoArchitect license that allows the planning of patterning sequences on a desktop PC.